Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCCTGAGAACCCTACACACCATC[A/G]TCACTCCATATGTTTATTAAATGAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Camk4 PubMed Links |
Camk4 - calcium/calmodulin-dependent protein kinase IV | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_009793.3 | Intron | NP_033923.2 |