Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Gabra3 PubMed Links |
Gabra3 - gamma-aminobutyric acid (GABA) A receptor, subunit alpha 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008067.4 | Intron | NP_032093.3 |
Mir105 - microRNA 105 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Mir767 - microRNA 767 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |