Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAAATTTGGGATGGTAATACTAG[C/T]ATGTTCAGGAGCATTTCACAATTTA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Pkhd1 PubMed Links |
Pkhd1 - polycystic kidney and hepatic disease 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153179.3 | Intron | NP_694819.2 |