Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCAATTGCCATTCCATGTCACATT[C/T]CATATAGTATGTAACTAGTGGGATG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cdh18 PubMed Links |
Cdh18 - cadherin 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081299.1 | Intron | NP_001074768.1 |