Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTAGGTGACCGTGGGCTGTGTT[A/G]AATTGATACTTAAAGCTAAGTAGGA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ttc7b PubMed Links |
Ttc7b - tetratricopeptide repeat domain 7B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033213.1 | Intron | NP_001028385.1 |