Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCATCTATAGCAGCATTTGCTA[C/T]TCAAAGCTGTCACATGCATTTTAGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Sdhaf4 PubMed Links |
Sdhaf4 - succinate dehydrogenase complex assembly factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_026503.3 | 480 | UTR 3 | NP_080779.2 |