Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATCAGATGCTCTGGAATACTAAT[A/C]AGATATACTGTGATAGCTGTGAATA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cdh12 PubMed Links |
Cdh12 - cadherin 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008420.2 | Intron | NP_001008420.1 |