Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAACCGGTTGTGCTCAGCAATAA[T/C]TCTGTCCAGCCTGGACACTTTCTCC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Syne1 PubMed Links |
Syne1 - spectrin repeat containing, nuclear envelope 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079686.1 | Intron | NP_001073154.1 | ||||
NM_022027.2 | Intron | NP_071310.2 | ||||
NM_153399.2 | Intron | NP_700448.2 |