Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCAGAGGGGTGGAGCATCTGGAG[C/G]CTGTGAGACTCTAGTGGGGAGGTGA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Elfn1 PubMed Links |
Elfn1 - leucine rich repeat and fibronectin type III, extracellular 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175522.3 | 3596 | UTR 3 | NP_780731.1 |