Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGATGTAATTATAGGTTATACATA[C/T]ACACTGTTCATTTCTGTAGGGAAAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Reln PubMed Links |
Reln - reelin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_011261.2 | Intron | NP_035391.2 |