Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGATTTCTGTCTTCGACTGTCAA[G/T]AGTCCCTTCTCGAAGCAAGTGTCCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ifih1 PubMed Links |
Ifih1 - interferon induced with helicase C domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164477.1 | 754 | Silent Mutation | NP_001157949.1 | |||
NM_027835.3 | 754 | Silent Mutation | NP_082111.2 |