Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGTGTTTATGGGTTTGTGTTGTT[A/G]GAAATGTCAGTTTCAGGTTTTTTCC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Plcl2 PubMed Links |
Plcl2 - phospholipase C-like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013880.3 | Intron | NP_038908.2 |