Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Abra PubMed Links |
Abra - actin-binding Rho activating protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175456.4 | 439 | Silent Mutation | NP_780665.1 |
Oxr1 - oxidation resistance 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |