Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTCTCAGACCAACGTGATTGGCC[A/G]ACCCTTTCCAGGGTCCTTGTGCAAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Slc38a11 PubMed Links |
Slc38a11 - solute carrier family 38, member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177074.2 | Intron | NP_796048.2 |