Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTGGCTTTAATAGCCCTTGACAT[G/T]TCTTGCTGAATGTTTACAAACTGAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Scn3a PubMed Links |
Scn3a - sodium channel, voltage-gated, type III, alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018732.3 | Intron | NP_061202.3 |