Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTAAAAGAAGGTTAATGGGTTTA[A/G]TGGTAGCTAGCTCTCCATCAACCCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Scn7a PubMed Links |
Scn7a - sodium channel, voltage-gated, type VII, alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_009135.2 | Intron | NP_033161.2 |