Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ckmt2 PubMed Links |
Ckmt2 - creatine kinase, mitochondrial 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198415.2 | Intron | NP_940807.1 |
Rasgrf2 - RAS protein-specific guanine nucleotide-releasing factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |