Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Lrg1 PubMed Links |
Lrg1 - leucine-rich alpha-2-glycoprotein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Plin4 - perilipin 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Plin5 - perilipin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077348.1 | 61 | UTR 5 | NP_001070816.1 | |||
NM_025874.3 | 61 | Intron | NP_080150.2 |
Sema6b - sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |