Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAACTCCCAAGAGCAAGGCCCAC[G/T]CGGAGAAGGCACGCCATGCCACTTA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Rab2a PubMed Links |
Rab2a - RAB2A, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021518.3 | 1711 | UTR 3 | NP_067493.1 |