Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAACATGTGGCTCACGAAGGTGTC[A/G]GGACTTTGGAGAATGGTGTCAAGAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Fbxw11 PubMed Links |
Fbxw11 - F-box and WD-40 domain protein 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271347.1 | Intron | NP_001258276.1 | ||||
NM_001271348.1 | Intron | NP_001258277.1 | ||||
NM_001271349.1 | Intron | NP_001258278.1 | ||||
NM_134015.3 | Intron | NP_598776.1 |