Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGCTTCTGGATTTCCACCAGCAC[A/G]GATGCGTCAAGCTGCTCGTGCAGAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Rgs22 PubMed Links |
Rgs22 - regulator of G-protein signalling 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195748.1 | 2874 | Silent Mutation | NP_001182677.1 |