Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCATTTATTGTTATAGTAACAGA[A/G]CTTCAACTGCACATAGCCTGGCTGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Itga4 PubMed Links |
Itga4 - integrin alpha 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_010576.3 | Intron | NP_034706.3 |