Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTGCTGCTCCTTATTTATTGTAA[C/T]GTGCTGCATGGAACATATTTATTTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Dync1i1 PubMed Links |
Dync1i1 - dynein cytoplasmic 1 intermediate chain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001191023.1 | 2679 | UTR 3 | NP_001177952.1 | |||
NM_001191025.1 | 2679 | UTR 3 | NP_001177954.1 | |||
NM_001191026.1 | 2679 | UTR 3 | NP_001177955.1 | |||
NM_001191027.1 | 2679 | UTR 3 | NP_001177956.1 | |||
NM_010063.4 | 2679 | UTR 3 | NP_034193.2 |