Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAACACACTCGGTGAGCCTCCGGC[A/C]TGAGCAGGCAGGGCGGGGTCCCGCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Atxn7 PubMed Links |
Atxn7 - ataxin 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139227.4 | 2526 | Missense Mutation | NP_631973.3 |