Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Gm16758 PubMed Links |
Gm16758 - predicted gene, 16758 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Mdga1 - MAM domain containing glycosylphosphatidylinositol anchor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081160.1 | Intron | NP_001074629.1 |