Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAGTAACAACTCTCATACTCTAC[A/G]GCTGTTTCCCAGTATCTAGGAATCC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
A430090L17Rik PubMed Links |
A430090L17Rik - RIKEN cDNA A430090L17 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Arl15 - ADP-ribosylation factor-like 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172595.4 | Intron | NP_766183.1 |