Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTAAAATGTAAATTCTACTCATT[A/C]TATTTCCTTCTATCAAGAATTCACT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ddx4 PubMed Links |
Ddx4 - DEAD (Asp-Glu-Ala-Asp) box polypeptide 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145885.1 | Intron | NP_001139357.1 | ||||
NM_010029.2 | Intron | NP_034159.1 |