Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGTGTACTGCTCAGAGGCAAGG[A/G]TTGCCTTGATTGCTTGATAATTTCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Lypla1 PubMed Links |
Lypla1 - lysophospholipase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008866.2 | Intron | NP_032892.1 |