Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Agpat9 PubMed Links |
Agpat9 - 1-acylglycerol-3-phosphate O-acyltransferase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172715.3 | 241 | UTR 5 | NP_766303.1 |
Fam175a - family with sequence similarity 175, member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |