Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCGAGAGACCGCAACTCAGAGAAC[C/T]GCAGTTTCTCAGCTTGAAAAAGACA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Xkr7 PubMed Links |
Xkr7 - X Kell blood group precursor related family member 7 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011732.1 | Intron | NP_001011732.1 |