Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGGAGTGTTAGTGTTAAATTGT[A/G]TCTATTTGGGACGTTATTAAGCAGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Fam19a2 PubMed Links |
Fam19a2 - family with sequence similarity 19, member A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252387.1 | Intron | NP_001239316.1 | ||||
NM_182807.3 | Intron | NP_877959.2 |