Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATGTTTCTCTATGGCTCTGGCCC[C/T]GAATACCTCAATGAATTGTGACCTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Raly PubMed Links |
Raly - hnRNP-associated with lethal yellow | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001139511.1 | Intron | NP_001132983.1 | ||||
NM_001139512.1 | Intron | NP_001132984.1 | ||||
NM_001139513.1 | Intron | NP_001132985.1 | ||||
NM_023130.3 | Intron | NP_075619.2 |