Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATAACCATCAGAATGTGATGCCAA[C/G]TTCCATCACATGAACTTTGCCTGAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Slc25a17 PubMed Links |
Slc25a17 - solute carrier family 25 (mitochondrial carrier, peroxisomal membrane protein), member 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_011399.3 | Intron | NP_035529.1 |