Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cenpk PubMed Links |
Cenpk - centromere protein K | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Ppwd1 - peptidylprolyl isomerase domain and WD repeat containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172807.4 | Intron | NP_766395.2 |