Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Gpr182 PubMed Links |
Gpr182 - G protein-coupled receptor 182 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Zbtb39 - zinc finger and BTB domain containing 39 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198035.1 | 1064 | Missense Mutation | NP_932152.1 |