Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGAGTTGCAAAATATACACACTG[C/T]ATTGAAATACTAGTTCTAGTAAAGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
4930567K20Rik PubMed Links |
4930567K20Rik - RIKEN cDNA 4930567K20 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Grm1 - glutamate receptor, metabotropic 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114333.2 | Intron | NP_001107805.1 | ||||
NM_016976.3 | Intron | NP_058672.1 |