Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGAAATCATGCTATTCTTACCAA[A/G]TCCCTGACACTTCCTTCAACAAGTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Kcnb2 PubMed Links |
Kcnb2 - potassium voltage gated channel, Shab-related subfamily, member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098528.2 | Intron | NP_001091998.1 |