Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Mir6942 PubMed Links |
Mir6942 - microRNA 6942 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Zkscan3 - zinc finger with KRAB and SCAN domains 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145778.1 | 716 | Missense Mutation | NP_001139250.1 | |||
NM_023685.4 | 716 | Missense Mutation | NP_076174.3 |