Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATGAATATCAGACTTTTAAAGAA[A/G]CAAAAGCAAAGTGTCACATTAAGGA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Exoc4 PubMed Links |
Exoc4 - exocyst complex component 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_009148.3 | Intron | NP_033174.2 |