Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCATCCTAGGAGCGCCTGCCCACT[C/T]CCGTCACCCTTCAATCTCCCATTCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Grm8 PubMed Links |
Grm8 - glutamate receptor, metabotropic 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008174.2 | Intron | NP_032200.2 |