Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGAAATTTTAAGATTATTTGAAT[C/T]AAGAAATGAAAACTAGATTCCATAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Pak7 PubMed Links |
Pak7 - p21 protein (Cdc42/Rac)-activated kinase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172858.2 | Intron | NP_766446.2 |