Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAGCTTAAAAATAAATACACACC[C/T]AAATTGCTGAGATTAATTGTAACCC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Nr3c1 PubMed Links |
Nr3c1 - nuclear receptor subfamily 3, group C, member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008173.3 | Intron | NP_032199.3 |