Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCATGGTAAGTTGTCAGCCATCC[A/G]CTGCCAAATGAAAAAGGAAGATGGA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Vps41 PubMed Links |
Vps41 - vacuolar protein sorting 41 (yeast) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172120.4 | Intron | NP_742118.3 |