Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGTTTATATCCCCTTTGCAAGAA[A/G]CACTTCTAAAAAGCACTCGCCACTT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ccdc14 PubMed Links |
Ccdc14 - coiled-coil domain containing 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172824.3 | 2345 | Missense Mutation | NP_766412.2 |