Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGGTGCAAGGGTGAGAGCTAGCA[G/T]ATACAATATGGAGTTCTGAGAGCAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Trappc9 PubMed Links |
Trappc9 - trafficking protein particle complex 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164641.1 | Intron | NP_001158113.1 | ||||
NM_001164642.1 | Intron | NP_001158114.1 | ||||
NM_001164643.1 | Intron | NP_001158115.1 | ||||
NM_029640.2 | Intron | NP_083916.1 | ||||
NM_180662.2 | Intron | NP_850993.2 |