Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGAAGAGAGAAAAAGAATGGCCA[A/G]GTCTCCTGTAAGCCACAGAAGCCAC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Gpr156 PubMed Links |
Gpr156 - G protein-coupled receptor 156 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153394.2 | Intron | NP_700443.2 |