Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGTCAATGGCTAACCCAGAAGAT[A/G]TGTATTAATGTGTCAAAAAGAATCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Smg6 PubMed Links |
Smg6 - Smg-6 homolog, nonsense mediated mRNA decay factor (C. elegans) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002764.1 | Intron | NP_001002764.1 |