Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGCTGCAATGGGGCCTTTAAACA[C/T]GGCCTTCTAAACTGGCTGACTGTGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Usp43 PubMed Links |
Usp43 - ubiquitin specific peptidase 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173754.4 | Intron | NP_776115.2 |