Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Dennd4c PubMed Links |
Dennd4c - DENN/MADD domain containing 4C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_184088.1 | Intron | NP_908976.1 |
Gm6521 - predicted gene 6521 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |