Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAGTGGCATCTAAGCAGCTGCCT[A/G]ACATTTGCAGACCAGATTTCTATGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cnih3 PubMed Links |
Cnih3 - cornichon homolog 3 (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160211.1 | Intron | NP_001153683.1 | ||||
NM_001160212.1 | Intron | NP_001153684.1 | ||||
NM_028408.3 | Intron | NP_082684.1 |